Learn More
Medchemexpress LLC GTP-14564 | 34823-86-4 | MFCD01416326 | 99.9% | 234.25 g·mol⁻¹ | C15H10N2O | 5 MG
SDP
Catalog No. 5000263862
Encompass_Preferred
GTP-14564 is a small-molecule tyrosine kinase inhibitor selective for internal tandem duplication (ITD) mutant and wild-type FLT3. It inhibits FLT3 ligand-dependent proliferation of Ba/F3 leukemia cells and is used as a research tool to study FLT3-driven signaling and leukemogenesis. Chemical formula C15H10N2O; molecular weight 234.25 g·mol⁻¹; reported purity 99.91%.
  • Targets ITD and wild-type FLT3.
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 cells.
  • High reported purity suitable for biochemical assays.
  • Compact quantity suitable for screening and small-scale studies.
  • Documented in chemical databases and literature for FLT3 research.
Catalog No. 50-002-63862 Supplier Medchemexpress LLC Supplier No. HY1100265MG
GTP-14564 is a small-molecule tyrosine kinase inhibitor selective for internal tandem duplication (ITD) mutant and wild-type FLT3. It inhibits FLT3 ligand-dependent proliferation of Ba/F3 leukemia cells and is used as a research tool to study FLT3-driven signaling and leukemogenesis. Chemical formula C15H10N2O; molecular weight 234.25 g·mol⁻¹; reported purity 99.91%.
  • Targets ITD and wild-type FLT3.
  • Inhibits FLT3 ligand-dependent growth in Ba/F3 cells.
  • High reported purity suitable for biochemical assays.
  • Compact quantity suitable for screening and small-scale studies.
  • Documented in chemical databases and literature for FLT3 research.