Learn More
American Radiolabeled Chemicals Inc GTP-8-3H 250UC
SDP
Catalog No. 5090104307
Encompass
Guanosine 5'-8-3H triphosphate tetrasodium salt 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 5-15 Ci/mmol, M.W. 611.2, Shipped in Dry Ice, (GTP)
Catalog No. 50-901-04307 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 0166250UC
Guanosine 5'-8-3H triphosphate tetrasodium salt 250uCi in ethanol:water (1:1), 1 mCi/ml, S.A. 5-15 Ci/mmol, M.W. 611.2, Shipped in Dry Ice, (GTP)