Learn More
American Radiolabeled Chemicals Inc GTP A-33P 1LC
SDP
Catalog No. 50901164
Encompass
Guanosine 5'-a-33P triphosphate tetra (triethylammonium) salt 1mCi in 50mM tricine (pH 7.6), 10 mCi/ml, S.A. 3000 Ci/mmol, M.W. 525.18, Shipped w/o dry ice, store at 4deg.C
Catalog No. 50-901-164 Supplier American Radiolabeled Chemicals Inc Supplier No. ARP 01551MC
Add to Cart
Add to Cart
Request a Quote
Guanosine 5'-a-33P triphosphate tetra (triethylammonium) salt 1mCi in 50mM tricine (pH 7.6), 10 mCi/ml, S.A. 3000 Ci/mmol, M.W. 525.18, Shipped w/o dry ice, store at 4deg.C