Learn More
Immunology Consultants GTP-Binding Nuclear Protein (RAN) (Host Cell Protein) ELISA Kit CHO (Chinese Hamster Ovary)
SDP
Catalog No. 5000435507
Encompass_Preferred
This ELISA kit enables quantitative detection of RAN, a GTP‑binding nuclear transport protein released as a CHO host cell impurity. RAN plays a key role in nucleocytoplasmic trafficking, and its presence in biologics can indicate incomplete purification. The assay provides high sensitivity and strong specificity for monitoring RAN across upstream and downstream samples. With a pre‑coated microplate and optimized reagents, E‑65RAN supports regulatory‑aligned HCP profiling, process validation, and impurity clearance studies.
Catalog No. 50-004-35507 Supplier Immunology Consultants Supplier No. E65RAN
This ELISA kit enables quantitative detection of RAN, a GTP‑binding nuclear transport protein released as a CHO host cell impurity. RAN plays a key role in nucleocytoplasmic trafficking, and its presence in biologics can indicate incomplete purification. The assay provides high sensitivity and strong specificity for monitoring RAN across upstream and downstream samples. With a pre‑coated microplate and optimized reagents, E‑65RAN supports regulatory‑aligned HCP profiling, process validation, and impurity clearance studies.