Learn More
Apexbio Technology LLC GTP-Binding Protein Fragment, G alpha 1mg
SDP
Catalog No. 5000092965
Encompass_Preferred
GTP-Binding Protein Fragment G alpha is a peptide fragment derived from the amino-terminal region of the alpha subunit of GTP-binding proteins It is designed to aid in elucidating the mechanisms of membrane association and -subunit interactions in cell signaling pathways GTP-Binding Protein Fragment G alpha exerts its biological activity by retaining membrane-binding properties independent of the nucleotide-binding state anchoring the protein to the cytoplasmic membrane surface This peptide fragment serves as an experimental tool to study membrane-association mechanisms and signal transduction pathways in biochemical and biomedical research Based on these properties GTP-Binding Protein Fragment G alpha holds research potential in investigating the molecular basis of G protein-mediated signal transduction and protein-membrane interactions
Catalog No. 50-000-92965 Supplier Apexbio Technology LLC Supplier No. A10221
GTP-Binding Protein Fragment G alpha is a peptide fragment derived from the amino-terminal region of the alpha subunit of GTP-binding proteins It is designed to aid in elucidating the mechanisms of membrane association and -subunit interactions in cell signaling pathways GTP-Binding Protein Fragment G alpha exerts its biological activity by retaining membrane-binding properties independent of the nucleotide-binding state anchoring the protein to the cytoplasmic membrane surface This peptide fragment serves as an experimental tool to study membrane-association mechanisms and signal transduction pathways in biochemical and biomedical research Based on these properties GTP-Binding Protein Fragment G alpha holds research potential in investigating the molecular basis of G protein-mediated signal transduction and protein-membrane interactions