Learn More
Enzo Life Sciences Guanosine 5'-O-(3-thiotriphosphate) . tetralithium salt (5mg). CAS: 94825-44-2
SDP
Supplier:  Enzo Life Sciences ALX480056M005
Encompass_Preferred
A nonhydrolyzable analog of guanosine 5’-O-triphosphate (GTP). Stimulates inositol trisphosphate formation in membranes. Alternative name: GTP-gamma-S . 4Li. Purity: ≥85% (HPLC). Solubility: Soluble in water (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9313