Learn More
Bioworld GTP SEPAROPORE
Catalog No. 50197834
Encompass
This item has a minimum order quantity of 2 per supplier requirements. Specifications: Ligand: Guanosine 5\'-Triphosphate (GTP); Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow specifications: 70-140 cm/h...
Catalog No. 50-197-834 Supplier Bioworld Supplier No. 201810661
This item has a minimum order quantity of 2 per supplier requirements. Specifications: Ligand: Guanosine 5\'-Triphosphate (GTP); Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow specifications: 70-140 cm/h...