Learn More
Bioworld GTP SEPAROPORE
Supplier:  Bioworld 201810663
Encompass
Specifications: Ligand: Guanosine 5\'-Triphosphate (GTP); Particle size (range): 50-175m; Molecular Weight range: 6x104 - 2x107; Matrix: Separopore 4B-CL (Crosslinked Agarose 4% Beads); pH stability: 4-9; Flow specifications: 70-140 cm/h...
Catalog No. 50-197-836