Learn More
Aviva Systems Biology Corp GTPBP10 Antibody - middle region (ARP42921_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42921_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTPBP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human GTPBP10. Alternate names: DKFZP686A10121, DKFZp686A10121, FLJ38242, MGC104191, ObgH2, UG0751c10ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58362