Learn More
Aviva Systems Biology Corp GTPBP2 Antibody - C-terminal region (ARP42539_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42539_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTPBP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GTPBP2. Alternate names: MGC74725ApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-58259