Learn More
Aviva Systems Biology Corp GTPBP9 Antibody - N-terminal region (ARP40745_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40745_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against GTPBP9. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human GTPBP9. Alternate names: DOC45, GBP45, GTBP9, GTPBP9, PTD004ApplicationWB. Species reactivity: Cow, Human, Mouse, Rat
Catalog No. 50-014-57606