Learn More
Aviva Systems Biology Corp GTPBP9 Antibody - N-terminal region (ARP40744_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40744_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GTPBP9. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human GTPBP9. Alternate names: DOC45, GBP45, GTBP9, GTPBP9, PTD004ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53233