Learn More
Selleck Chemical LLC GTS-21 dihydrochloride
SDP
Catalog No. 503135784
Encompass_Preferred
GTS-21 dihydrochloride(DMXB-A DMBX-anabaseine) is a selective agonist of alpha7 nicotinic acetylcholine receptor ( 7-nAChR) and exhibits anti inflammatory and cognition enhancing activities GTS-21 dihydrochloride is also an antagonist of 4 2 with a Ki value of 20 nM for human 4 2 and 5-HT3A receptor with an IC50 value of 3 1 M
Catalog No. 50-313-5784 Supplier Selleck Chemical LLC Supplier No. E187625MG
GTS-21 dihydrochloride(DMXB-A DMBX-anabaseine) is a selective agonist of alpha7 nicotinic acetylcholine receptor ( 7-nAChR) and exhibits anti inflammatory and cognition enhancing activities GTS-21 dihydrochloride is also an antagonist of 4 2 with a Ki value of 20 nM for human 4 2 and 5-HT3A receptor with an IC50 value of 3 1 M