Learn More
Medchemexpress LLC HY-14564A 5mg Medchemexpress, GTS-21 (dihydrochloride) CAS:156223-05-1 Purity:>98%
SDP
Catalog No. 502081613
Encompass_Preferred
Medchemexpress, HY-14564A 5mg GTS-21 (dihydrochloride) CAS:156223-05-1 GTS-21 dihydrochloride is a selective alpha7 nicotinic acetylcholine receptor (α7-nAChR) agonist with anti‑inflammatory and cognition‑enhancing activities. GTS-21 dihydrochloride is also a α4β2 (Ki=20 nM for humanα4β2) and 5-HT3A receptor (IC50=3.1 μM) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1613 Supplier Medchemexpress LLC Supplier No. HY14564A5MG
Medchemexpress, HY-14564A 5mg GTS-21 (dihydrochloride) CAS:156223-05-1 GTS-21 dihydrochloride is a selective alpha7 nicotinic acetylcholine receptor (α7-nAChR) agonist with anti‑inflammatory and cognition‑enhancing activities. GTS-21 dihydrochloride is also a α4β2 (Ki=20 nM for humanα4β2) and 5-HT3A receptor (IC50=3.1 μM) antagonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.