Learn More
Selleck Chemical LLC GTS-21 dihydrochloride
Supplier: Selleck Chemical LLC E18765MG
GTS-21 dihydrochloride(DMXB-A DMBX-anabaseine) is a selective agonist of alpha7 nicotinic acetylcholine receptor ( 7-nAChR) and exhibits anti inflammatory and cognition enhancing activities GTS-21 dihydrochloride is also an antagonist of 4 2 with a Ki value of 20 nM for human 4 2 and 5-HT3A receptor with an IC50 value of 3 1 M