Learn More
Cayman Chemical GTS-21 (hydrochloride), Purity: ≥98%, CAS: 156223-05-1, 50 mg
SDP
Catalog No. 502964009
Encompass_Preferred
An α7-containing nAChR agonist; activates human α7-containing nAChRs to 60% of the levels generated by endogenous ligand acetylcholine; has no activity at α2-, α3-, and α4-containing nAChRs at a concentration of 100 μM; stimulates dopamine release from rat striatal slices (EC50 = 10 μM); decreases delay in the delayed matching-to-sample task in monkeys; suppresses amnesia and neuronal cell death in a gerbil model of ischemia-reperfusion injury
Catalog No. 50-296-4009 Supplier Cayman Chemical Supplier No. 2157050 MG
An α7-containing nAChR agonist; activates human α7-containing nAChRs to 60% of the levels generated by endogenous ligand acetylcholine; has no activity at α2-, α3-, and α4-containing nAChRs at a concentration of 100 μM; stimulates dopamine release from rat striatal slices (EC50 = 10 μM); decreases delay in the delayed matching-to-sample task in monkeys; suppresses amnesia and neuronal cell death in a gerbil model of ischemia-reperfusion injury