Learn More
OHAUS GTS Adhering Mat Platform
Catalog No. 01804347 Shop All OHAUS Products
Change view
Click to view available options
Product Type:
Shaker Attachment
Catalog No. Product Type
01-804-347 Shaker Attachment
1 options
Catalog No. 01-804-347 Supplier OHAUS Supplier No. 83041277
Interchangeable Adhering Mat platform.
  • For use on all Genie Temp-Shaker models.
  • Remove existing platform and install this one with no tools required.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Shaker Attachment