Learn More
Waters Corp SEC Column, 450 Å, 2.5 μm, 4.6 x 300 mm
Catalog No. NC2389556 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column offers high-resolution, rapid size variant separations for AAVs and nucleic acids, essential for gene therapy. Its 450 Å pore size resolves monomers and aggregates. MaxPeak High Performance Surfaces mitigate secondary interactions for robust methods. The 2.5 μm particles ensure industry-leading resolution.
  • MaxPeak high performance surfaces minimize secondary interactions with hardware.
  • BEH technology particle ensures rugged, inert performance for size variant analyses.
  • Provides industry-leading multi-angle light scattering (MALS) sensitivity.
  • Supports method flexibility and robustness across diverse buffers.
  • Manufactured in a cGMP, ISO 9001 certified plant with stringent QC.
  • Enables fast, reliable gene therapeutic analysis and enhanced aggregate characterization.
Catalog No. NC2389556 Supplier Waters Corp Supplier No. 186010585
Add to Cart
Add to Cart
This column offers high-resolution, rapid size variant separations for AAVs and nucleic acids, essential for gene therapy. Its 450 Å pore size resolves monomers and aggregates. MaxPeak High Performance Surfaces mitigate secondary interactions for robust methods. The 2.5 μm particles ensure industry-leading resolution.
  • MaxPeak high performance surfaces minimize secondary interactions with hardware.
  • BEH technology particle ensures rugged, inert performance for size variant analyses.
  • Provides industry-leading multi-angle light scattering (MALS) sensitivity.
  • Supports method flexibility and robustness across diverse buffers.
  • Manufactured in a cGMP, ISO 9001 certified plant with stringent QC.
  • Enables fast, reliable gene therapeutic analysis and enhanced aggregate characterization.