Learn More
Waters Corp GTxResolve Column, 1000 Å, 3 μm, 7.8 x 150 mm
Catalog No. NC2885817 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This column is designed for high-recovery analyses of Gene Therapy (GTx) drugs with diameters ranging from 200 to 800 Å, including mRNA, LNPs, and viral vectors. It features low adsorption MaxPeak HPS hardware and innovative bridged ethylene HO-PEO particles.
  • Long column lifetimes
  • Minimal secondary interactions
  • Enhanced MALS sensitivity
  • Facilitates the development of platform multiattribute SEC assays for assessing mRNA integrity and LNP formulations
  • Provides improved detection of ejected transgenes and aggregates associated with viral vectors
Catalog No. NC2885817 Supplier Waters Corp Supplier No. 186010737
This column is designed for high-recovery analyses of Gene Therapy (GTx) drugs with diameters ranging from 200 to 800 Å, including mRNA, LNPs, and viral vectors. It features low adsorption MaxPeak HPS hardware and innovative bridged ethylene HO-PEO particles.
  • Long column lifetimes
  • Minimal secondary interactions
  • Enhanced MALS sensitivity
  • Facilitates the development of platform multiattribute SEC assays for assessing mRNA integrity and LNP formulations
  • Provides improved detection of ejected transgenes and aggregates associated with viral vectors