Learn More
Waters Corp Column, SEC, 1000 Å, 3 μm, 7.8 x 300 mm
Supplier: Waters Corp 186010738
This SEC column features low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles. It is designed to support high recovery analyses of 200 to 800Å diameter gene therapeutics drugs, including mRNA, LNPs, and viral vectors. This column technology offers long column lifetimes, reduced secondary interactions, and enhanced MALS sensitivity. It facilitates the development of platform multiattribute SEC assays for mRNA integrity, LNP formulations analyses, and improved detection of ejected transgenes and viral vector-related aggregates.
- Pore size: 1000 Å
- Particle size: 3 μm
- Inner diameter: 7.8 mm
- Length: 300 mm
- pH range: 2.5 to 8 pH
- Maximum temperature: 60 °C
- Maximum pressure: 3750 psi (258 Bar)
- Compatible with UHPLC, UPLC, and HPLC systems
- Particle substrate: Silica
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.