Learn More
Waters Corp Column, 1000 Å, 3 μm, 4.6 mm ID, 150 mm Length
Catalog No. NC2885815 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GTxResolve Premier SEC 1000 Å, 3 μm, 4.6 x 150 mm Column is designed for high recovery analyses of 200 to 800Å diameter GTx drugs, utilizing low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Supports high recovery analysis of 200 to 800Å diameter GTx drugs.
  • Provides long column lifetimes, low secondary interactions, and improved MALS sensitivity.
  • High strength silica packing with PEO bonding and bridged ethylene hybrid crosslinks.
  • Offers superior chemical inertness, increased pH resilience, and reduced chemical noise.
  • cGMP, ISO 9001 certified manufacturing with rigorous QC.
  • Designed for method flexibility and robustness across diverse buffers and organic modifiers.
Catalog No. NC2885815 Supplier Waters Corp Supplier No. 186010735
The GTxResolve Premier SEC 1000 Å, 3 μm, 4.6 x 150 mm Column is designed for high recovery analyses of 200 to 800Å diameter GTx drugs, utilizing low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Supports high recovery analysis of 200 to 800Å diameter GTx drugs.
  • Provides long column lifetimes, low secondary interactions, and improved MALS sensitivity.
  • High strength silica packing with PEO bonding and bridged ethylene hybrid crosslinks.
  • Offers superior chemical inertness, increased pH resilience, and reduced chemical noise.
  • cGMP, ISO 9001 certified manufacturing with rigorous QC.
  • Designed for method flexibility and robustness across diverse buffers and organic modifiers.