Learn More
Waters Corp HPLC Column, 1000 Å, 3 μm, 4.6 mm, 300 mm
Catalog No. NC3862449 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GTxResolve Premier SEC 1000 Å, 3 μm, 4.6 x 300 mm Column is engineered with MaxPeak HPS hardware and HO-PEO particles for high recovery analysis of GTx drugs. It provides long column lifetimes and improved MALS sensitivity.
  • Manufactured with low adsorption MaxPeak HPS hardware.
  • Uses novel bridged ethylene HO-PEO particles.
  • Supports high recovery analyses of 200 to 800Å diameter GTx drugs, such as mRNA, LNPs, and viral vectors.
  • Provides long column lifetimes.
  • Offers low secondary interactions.
  • Improved MALS sensitivity.
  • Facilitates the development of platform multiattribute SEC assays for mRNA integrity and LNP formulations analyses.
  • Improved detection of ejected transgenes and aggregates related to viral vectors.
Catalog No. NC3862449 Supplier Waters Corp Supplier No. 176006043
The GTxResolve Premier SEC 1000 Å, 3 μm, 4.6 x 300 mm Column is engineered with MaxPeak HPS hardware and HO-PEO particles for high recovery analysis of GTx drugs. It provides long column lifetimes and improved MALS sensitivity.
  • Manufactured with low adsorption MaxPeak HPS hardware.
  • Uses novel bridged ethylene HO-PEO particles.
  • Supports high recovery analyses of 200 to 800Å diameter GTx drugs, such as mRNA, LNPs, and viral vectors.
  • Provides long column lifetimes.
  • Offers low secondary interactions.
  • Improved MALS sensitivity.
  • Facilitates the development of platform multiattribute SEC assays for mRNA integrity and LNP formulations analyses.
  • Improved detection of ejected transgenes and aggregates related to viral vectors.