Learn More
Waters Corp GTxResolve Premier SEC Guard, 1000Å, 3 μm, 4.6 x 30 mm
Catalog No. NC2885814 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The GTxResolve Premier SEC 1000Å, 3 μm, 4.6 x 30 mm Guard is manufactured with low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles. It is designed to support high recovery analyses of 200 to 800Å diameter GTx drugs, including mRNA, LNPs, and viral vectors. This optimized 4.6 x 30 mm Guard is intended for use with both 4.6 and 7.8 mm ID analytical columns to ensure long column lifetimes.
  • Features low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Designed for high recovery analyses of GTx drugs (200 to 800Å), including mRNA, LNPs, and viral vectors.
  • Extends analytical column lifetime by trapping particulates; cGMP, ISO 9001 certified manufacturing with rigorous QC.
  • Minimizes secondary interactions, offering broad method flexibility across buffers (pH 2.5–8.5, 4°C to 50°C).
Catalog No. NC2885814 Supplier Waters Corp Supplier No. 186010733
The GTxResolve Premier SEC 1000Å, 3 μm, 4.6 x 30 mm Guard is manufactured with low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles. It is designed to support high recovery analyses of 200 to 800Å diameter GTx drugs, including mRNA, LNPs, and viral vectors. This optimized 4.6 x 30 mm Guard is intended for use with both 4.6 and 7.8 mm ID analytical columns to ensure long column lifetimes.
  • Features low adsorption MaxPeak HPS hardware and novel bridged ethylene HO-PEO particles.
  • Designed for high recovery analyses of GTx drugs (200 to 800Å), including mRNA, LNPs, and viral vectors.
  • Extends analytical column lifetime by trapping particulates; cGMP, ISO 9001 certified manufacturing with rigorous QC.
  • Minimizes secondary interactions, offering broad method flexibility across buffers (pH 2.5–8.5, 4°C to 50°C).