Learn More
TARGETMOL CHEMICALS INC GUACETISAL 5MG
SDP
Catalog No. 502581225
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Guacetisal is extracted from the esterification reaction of acetylsalicylic acid with guaiacol which can be used for research on the treatment of chronic bronchitis. purity: 98%
Catalog No. 50-258-1225 Supplier TARGETMOL CHEMICALS INC Supplier No. T154475MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Guacetisal is extracted from the esterification reaction of acetylsalicylic acid with guaiacol which can be used for research on the treatment of chronic bronchitis. purity: 98%