Learn More
Medchemexpress LLC HY-17477 5mg Medchemexpress, Guacetisal CAS:55482-89-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY174775MG
Encompass_Preferred
Medchemexpress, HY-17477 5mg Guacetisal CAS:55482-89-8 Guacetisal is obtained from the esterification of acetylsalicylic acid with guaiacol which has the potential for chronic bronchitis treatment extracted from patent CN 106866420 A. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1964