Learn More
Cayman Chemical Guaifenesin, Purity: ≥98%, CAS: 93-14-1, 250 g
SDP
Catalog No. 502985055
Encompass_Preferred
An expectorant; inhibits MUC5AC production, reduces mucus viscosity and elasticity, and increases the mucociliary transport rate of endogenous particles in primary human tracheobronchial epithelial cells; increases phenol red secretion in rats at 50 mg/kg
Catalog No. 50-298-5055 Supplier Cayman Chemical Supplier No. 21250250 G
An expectorant; inhibits MUC5AC production, reduces mucus viscosity and elasticity, and increases the mucociliary transport rate of endogenous particles in primary human tracheobronchial epithelial cells; increases phenol red secretion in rats at 50 mg/kg