Learn More
Cayman Chemical Guaifenesin, Purity: ≥98%, CAS: 93-14-1, 50 mg
SDP
Catalog No. 502950012
Encompass_Preferred
An analytical reference standard categorized as an expectorant; intended for use in analytical forensic applications
Catalog No. 50-295-0012 Supplier Cayman Chemical Supplier No. 3299850 MG
An analytical reference standard categorized as an expectorant; intended for use in analytical forensic applications