Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cole-Parmer Ashcroft BY2.5 2.5" Liquid Filled Pressure Gauge, 0 to 100 psi, Lower

Supplier:  Cole-Parmer 6842923

Encompass_Preferred

FEATURES

  • Available in 40mm, 50mm, 63mm and 100mm sizes
  • Stainless steel case and ring with plastic window
  • 1/8 NPT back connection available in 40mm size
  • 1/4 NPT lower and back connection available in 50mm and 63mm sizes
  • 1/4 NPT lower connection available in 100mm size
  • Dual scale dials with kPa in blue (outer scale); psi in black (inner scale); on white background
  • Panel mount kits available

Catalog No. 50-992-474


May include imposed supplier surcharges.
Add to Cart

Description

Description

Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.