Learn More
Cole-Parmer Ashcroft 1188AS4.5 4.5" Low-Pressure Brass Bellows Gauge 0 to 30" WC
Catalog No. 50992427 Shop All Cole-Parmer Products
Encompass_Preferred
41/2'' Low Pressure (from 10'' H2O to 10 psi) Bellows Gauge
Grade A (2-1-2% F.S. Accuracy)
With ranges as low as 10 inches of water vacuum, a variety of wetted parts and case materials and solid front case designs, these low pressure gauges are generally used on applications measuring air, liquid or gas.
Catalog No. 50-992-427 Supplier Cole-Parmer Supplier No. 6842750
41/2'' Low Pressure (from 10'' H2O to 10 psi) Bellows Gauge
Grade A (2-1-2% F.S. Accuracy)
With ranges as low as 10 inches of water vacuum, a variety of wetted parts and case materials and solid front case designs, these low pressure gauges are generally used on applications measuring air, liquid or gas.