Learn More
Cole-Parmer Ashcroft 1279AS4.5 4.5" DuraGauge Pressure Gauge 0 to 100 psi
Catalog No. 50992434 Shop All Cole-Parmer Products
Encompass_Preferred
This Pressure Gauge is the finest production gauge on the market for industrial use where a precise indication is required. The line offers a wide variety of case styles, Bourdon tubes, and pressure ranges to meet any conceivable application. The case style is a solid-front safety design for the gauge. Offered in 4-1/2'' size only, it is weatherproof; with hermetically-sealed or liquid-filled versions.
Catalog No. 50-992-434 Supplier Cole-Parmer Supplier No. 6842690
This Pressure Gauge is the finest production gauge on the market for industrial use where a precise indication is required. The line offers a wide variety of case styles, Bourdon tubes, and pressure ranges to meet any conceivable application. The case style is a solid-front safety design for the gauge. Offered in 4-1/2'' size only, it is weatherproof; with hermetically-sealed or liquid-filled versions.