Learn More
Cole-Parmer Ashcroft 10824.5 4.5" Precision Test Gauge 0 to 5000 psi
Catalog No. 50992451 Shop All Cole-Parmer Products
Encompass_Preferred
This new generation of test gauges to meet the exacting needs of instrument engineers and quality specialists. The new Ashcroft test gauge can be used as a transfer standard for periodically checking the accuracy of pressure instruments or as a laboratory instrument for research and test labs. The wealth of new features makes it an excellent test gauge for process applications where high accuracy in pressure measurement is essential. FEATURES
  • Precision accuracy ±0.25% full scale, ASME Grade 3A
  • Temperature compensated reduces ambient temperature errors to less than 0.005% per °F
  • External micro-span adjustment for ease in span calibration from 0-100% of scale
  • Hydraulically staked movement with PTFE coated gears and bearings improves stability
  • Precision dial and pointer results in excellent readability
Catalog No. 50-992-451 Supplier Cole-Parmer Supplier No. 6842533
Add to Cart
Add to Cart
This new generation of test gauges to meet the exacting needs of instrument engineers and quality specialists. The new Ashcroft test gauge can be used as a transfer standard for periodically checking the accuracy of pressure instruments or as a laboratory instrument for research and test labs. The wealth of new features makes it an excellent test gauge for process applications where high accuracy in pressure measurement is essential. FEATURES
  • Precision accuracy ±0.25% full scale, ASME Grade 3A
  • Temperature compensated reduces ambient temperature errors to less than 0.005% per °F
  • External micro-span adjustment for ease in span calibration from 0-100% of scale
  • Hydraulically staked movement with PTFE coated gears and bearings improves stability
  • Precision dial and pointer results in excellent readability