Learn More
Cole-Parmer Ashcroft EA14B 4.5" Contractor Pressure Gauge, 0 to 60 psi
Catalog No. 50992437 Shop All Cole-Parmer Products
Encompass_Preferred
  • 4-1/2" corrosion resistant stainless steel case and ring, plastic window
  • Accuracy is 1% full scale
  • Balanced adjustable black pointer, adjust screw in face of instrument
  • Phosphor bronze movement
  • Soft soldered phosphor bronze tube with brass socket; suitable for steam, water, air, oil, gas or any medium not corrosive to copper allows
  • White dial with bold black graduations and figures
  • 1/4 NPT bottom connection only
Catalog No. 50-992-437 Supplier Cole-Parmer Supplier No. 6842760
  • 4-1/2" corrosion resistant stainless steel case and ring, plastic window
  • Accuracy is 1% full scale
  • Balanced adjustable black pointer, adjust screw in face of instrument
  • Phosphor bronze movement
  • Soft soldered phosphor bronze tube with brass socket; suitable for steam, water, air, oil, gas or any medium not corrosive to copper allows
  • White dial with bold black graduations and figures
  • 1/4 NPT bottom connection only