Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiac Tincture, 1:60 in Alcohol, for Blood Detection, LabChem™

Catalog No. LC149151
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC149151 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC149151 Supplier LabChem, Inc. Supplier No. LC149151

Specifications

Chemical Name or Material Guaiac Tincture
Color Brown
Name Note 1:60 in Alcohol, for Blood Detection
CAS Max % 84.1, 4.92, 4.67, 4.67, 1.64
CAS 7732-18-5, 67-63-0, 67-56-1, 9000-29-7
UN Number UN1170
Solubility Information Soluble in water
Identification Passes Test
Packaging Poly Bottle
Quantity 500 mL
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.