Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiac Tincture 1:60, Ricca Chemical
SDP

Catalog No. 34604 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34604 120 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34604 Supplier Ricca Chemical Company Supplier No. 34604

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material Guaiac Tincture
CAS Min % 81.0, 5.93, 4.3, 4.0, 1.98
CAS Max % 85.0, 6.17, 4.48, 6.0, 2.06
CAS 7732-18-5, 67-63-0, 67-56-1, 9000-29-7
Concentration 1:60
Packaging Natural Poly Bottle
Color Colorless
Quantity 120 mL
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.