Learn More
Medchemexpress LLC Guaiacol | 90-05-1 | 124.14 | 100 G
SDP
Catalog No. 5000414879
Encompass_Preferred
Guaiacol, also known as 2-Methoxyphenol, is a phenolic compound exhibiting anti-inflammatory activity. It is known to inhibit LPS-stimulated COX-2 expression and NF-κB activation, with an IC50 for COX-2 and NF-κB. This compound also demonstrates antiproliferative activity against human U251MG and U87MG cells.
  • Inhibits LPS-stimulated NF-κB activation and COX-2 gene expression
  • Exhibits anti-inflammatory properties
  • Shows antiproliferative activity against U-251 and U-87MG human cell lines
  • Soluble in DMSO (≥ 100 mg/mL) and water (37.5 mg/mL)
Catalog No. 50-004-14879 Supplier Medchemexpress LLC Supplier No. HYN1380100G
Guaiacol, also known as 2-Methoxyphenol, is a phenolic compound exhibiting anti-inflammatory activity. It is known to inhibit LPS-stimulated COX-2 expression and NF-κB activation, with an IC50 for COX-2 and NF-κB. This compound also demonstrates antiproliferative activity against human U251MG and U87MG cells.
  • Inhibits LPS-stimulated NF-κB activation and COX-2 gene expression
  • Exhibits anti-inflammatory properties
  • Shows antiproliferative activity against U-251 and U-87MG human cell lines
  • Soluble in DMSO (≥ 100 mg/mL) and water (37.5 mg/mL)