Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Guaiacol Concentrate
SCI_ED

Catalog No. S37736 Shop All Aldon Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S37736 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S37736 Supplier Innovating Science™ Supplier No. IS3730
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guaiacol concentrate, 5 mL used in IS3713 AP Biology Investigation #13: Enzyme Activity

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Color Solid ≤82.4°F / Liquid ≥89.6°F
Boiling Point 205°C
Melting Point 28°C
Solubility Information Slightly soluble in water
Formula Weight 124.14
Percent Purity Concentrate
Grade Reagent
Odor Phenol-like
Packaging Amber Glass Bottle
Vapor Pressure 100 at 20°C
Vapor Density 4.27
pH 7.2
Quantity 5 mL
Flash Point 88.8°C (192°F) CC
Chemical Name or Material Guaiacol
Show More Show Less
EINECSNumber 201-964-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.