Learn More
Medchemexpress LLC Guaiacol-d3 | 74495-69-5 | 127.16 | 50 MG
SDP
Catalog No. 5000415107
Encompass_Preferred
Guaiacol-d3 is the deuterium labeled Guaiacol. Guaiacol, a phenolic compound, inhibits LPS-stimulated COX-2 expression and NF-κB activation. Guaiacol has an anti-inflammatory activity.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15107 Supplier Medchemexpress LLC Supplier No. HYN1380S150MG
Guaiacol-d3 is the deuterium labeled Guaiacol. Guaiacol, a phenolic compound, inhibits LPS-stimulated COX-2 expression and NF-κB activation. Guaiacol has an anti-inflammatory activity.
  • This compound can be used as a tracer.
  • This compound can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.