Learn More
Cosmo Bio Usa Inc Va-YSG Medium, low pH culture medium, 100 tubes, 2 mL per tube, for detecting *Alicyclobacillus acidoterrestris* in fruit juices
SDP
Catalog No. NC9002541
Encompass
Va-YSG Medium, low pH culture medium, 100 tubes, 2 mL per tube, for detecting *Alicyclobacillus acidoterrestris* in fruit juices
Catalog No. NC9002541 Supplier Cosmo Bio Usa Inc Supplier No. KYO08901
Va-YSG Medium, low pH culture medium, 100 tubes, 2 mL per tube, for detecting *Alicyclobacillus acidoterrestris* in fruit juices