Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiacol, Purified, 99%, Spectrum™ Chemical
SDP

Catalog No. 18603067 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-067 500 mL Amber Glass Bottle
18-603-066 100 mL Amber Glass Bottle
18-603-068 4 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-067 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. GU101500MLGL
Only null left
Add to Cart
Add to Cart

GU101, 90-05-1, C7H8O2

Spectrum™ Chemical Guaiacol, Purified is a natural organic compound that is biosynthesized by several different organisms. It is a yellow pleasant smelling oil derived most commonly from wood creosote. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Percent Purity 99%
Grade Purified
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.