Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiacol, Purified, 99%, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
4 L
Packaging:
Amber Glass Bottle
90-05-1
C7H8O2
124.14
LHGVFZTZFXWLCP-UHFFFAOYSA-N
2-methoxyphenol
COC1=CC=CC=C1O
90-05-1
C7H8O2
124.14
LHGVFZTZFXWLCP-UHFFFAOYSA-N
2-methoxyphenol
COC1=CC=CC=C1O

GU101, 90-05-1, C7H8O2

Spectrum™ Guaiacol, Purified is a natural organic compound that is biosynthesized by several different organisms. It is a yellow pleasant smelling oil derived most commonly from wood creosote. The Purified Grade describes chemicals of good quality where there are no official standards and is normally limited to inorganic chemicals.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 1
Percent Purity 99%