Learn More
Medchemexpress LLC Guaiacylglycerol-bet 500mg
SDP
Catalog No. 5000206388
Encompass_Preferred
Guaiacylglycerol-beta-guaiacyl ether is a model compound of lignin monomers that can be degraded by Bacillus amyloliquefaciens CotA[1]
Catalog No. 50-002-06388 Supplier Medchemexpress LLC Supplier No. HYW114954500MG
Guaiacylglycerol-beta-guaiacyl ether is a model compound of lignin monomers that can be degraded by Bacillus amyloliquefaciens CotA[1]