Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guaiacylglycerol-beta-guaiacyl Ether 97.0+%, TCI America™
SDP

Catalog No. G02331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02331G 1 g
G0233100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02331G Supplier TCI America Supplier No. G02331G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 7382-59-4
Molecular Formula C17H20O6
Molecular Weight (g/mol) 320.34
MDL Number MFCD00060144
InChI Key COXIJIHARCXZBW-UHFFFAOYNA-N
Synonym 1-(4-Hydroxy-3-methoxyphenyl)-2-(2-methoxyphenoxy)-1,3-propanediol
PubChem CID 6424189
ChEBI CHEBI:53650
IUPAC Name 4-[1-hydroxy-2-(hydroxymethoxy)-2-(2-methoxyphenyl)ethyl]-2-methoxyphenol
SMILES COC1=CC(=CC=C1O)C(O)C(OCO)C1=CC=CC=C1OC

Specifications

Melting Point 100°C
Color White
Quantity 1 g
Formula Weight 320.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Guaiacylglycerol-beta-guaiacyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.