Learn More
Medchemexpress LLC Guaiazulene (standard) | 489-84-9 | MFCD00003811 | 25 MG
SDP
Catalog No. 5000421593
Encompass_Preferred
Guaiazulene (Standard) is an analytical standard intended for research and analytical applications. It is a bicyclic sesquiterpene that can cross the blood-brain barrier and exhibits various biological activities, including anti-inflammatory, antioxidant, hepatoprotective, antibacterial, and anti-tumor properties. It is also used as a colorant in cosmetics. At high concentrations, Guaiazulene shows in vitro cytotoxicity to rat neuronal cells and N2a neuroblastoma cells.
  • Analytical standard for research applications
  • Reference standard supplied assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Applicable in HPLC, GC, and MS
Catalog No. 50-004-21593 Supplier Medchemexpress LLC Supplier No. 25MGHYN6951R
Guaiazulene (Standard) is an analytical standard intended for research and analytical applications. It is a bicyclic sesquiterpene that can cross the blood-brain barrier and exhibits various biological activities, including anti-inflammatory, antioxidant, hepatoprotective, antibacterial, and anti-tumor properties. It is also used as a colorant in cosmetics. At high concentrations, Guaiazulene shows in vitro cytotoxicity to rat neuronal cells and N2a neuroblastoma cells.
  • Analytical standard for research applications
  • Reference standard supplied assay
  • Used in qualitative research experiments
  • Used in quantitative research experiments
  • Used in methodological research experiments
  • Applicable in HPLC, GC, and MS