Learn More
Medchemexpress LLC HY-B0264 500mg Medchemexpress, Guaifenesin CAS:93-14-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0264500MG
Encompass_Preferred
Medchemexpress, HY-B0264 500mg Guaifenesin CAS:93-14-1 Guaifenesin (Guaiacol glyceryl ether), a constituent of guaiac resin from the wood of Guajacum officinale Linné, is an expectorant. Guaifenesin can alleviate cough discomfortby increasing sputum volume and decreasing its viscosity, thereby promoting effective cough. Guaifenesin also has narcotic effect. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2434