Learn More
Medchemexpress LLC Guaifenesin-d3 | 1189924-85-3 | 99.0% | 1 MG
SDP
Catalog No. 5000406513
Encompass_Preferred
Guaifenesin-d3 is the deuterium labeled Guaifenesin. Guaifenesin (Guaiacol glyceryl ether), a constituent of guaiac resin from the wood of Guajacum officinale Linné, is an expectorant. It can alleviate cough discomfort by increasing sputum volume and decreasing its viscosity, thereby promoting effective cough.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Derived from guaiac resin from the wood of Guajacum officinale Linné.
  • Helps alleviate cough discomfort by increasing sputum volume and decreasing its viscosity.
  • Promotes effective cough.
Catalog No. 50-004-06513 Supplier Medchemexpress LLC Supplier No. HYB0264S1MG
Guaifenesin-d3 is the deuterium labeled Guaifenesin. Guaifenesin (Guaiacol glyceryl ether), a constituent of guaiac resin from the wood of Guajacum officinale Linné, is an expectorant. It can alleviate cough discomfort by increasing sputum volume and decreasing its viscosity, thereby promoting effective cough.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Derived from guaiac resin from the wood of Guajacum officinale Linné.
  • Helps alleviate cough discomfort by increasing sputum volume and decreasing its viscosity.
  • Promotes effective cough.