Learn More
Medchemexpress LLC Guaifenesin-d3 | 1189924-85-3 | MFCD03428243 | 99.0% | 201.23 | C10H11D3O4 | 10 MG
SDP
Catalog No. 5000298219
Encompass_Preferred
Guaifenesin-d3 is the deuterium-labeled form of guaifenesin provided as a solid reference material for analytical applications. It is intended for research use as a tracer or internal standard to support quantitative NMR, GC-MS, and LC-MS measurements, delivering consistent calibration and accurate mass-based analyses.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity (99.0%) for reliable results.
  • Formula C10H11D3O4 and molecular weight 201.23 for accurate mass calculations.
  • Solid, white to off-white appearance for straightforward handling.
  • Stable storage: powder kept at -20°C retains stability for up to 3 years.
Catalog No. 50-002-98219 Supplier Medchemexpress LLC Supplier No. HYB0264S10MG
Guaifenesin-d3 is the deuterium-labeled form of guaifenesin provided as a solid reference material for analytical applications. It is intended for research use as a tracer or internal standard to support quantitative NMR, GC-MS, and LC-MS measurements, delivering consistent calibration and accurate mass-based analyses.
  • Deuterium-labeled internal standard for quantitative analysis.
  • High chemical purity (99.0%) for reliable results.
  • Formula C10H11D3O4 and molecular weight 201.23 for accurate mass calculations.
  • Solid, white to off-white appearance for straightforward handling.
  • Stable storage: powder kept at -20°C retains stability for up to 3 years.