Learn More
Cayman Chemical Guaifenesin-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 1189924-85-3, 1 mg
SDP
Catalog No. 502947165
Encompass_Preferred
An internal standard for the quantification of guaifenesin by GC- or LC-MS
Catalog No. 50-294-7165 Supplier Cayman Chemical Supplier No. 333681 MG
An internal standard for the quantification of guaifenesin by GC- or LC-MS