Learn More
Cayman Chemical GuaIfensIn-d3 500ug
SDP
Catalog No. 502947691
Encompass_Preferred
An internal standard for the quantification of guaifenesin by GC- or LC-MS
Catalog No. 50-294-7691 Supplier Cayman Chemical Supplier No. 33368500 UG
An internal standard for the quantification of guaifenesin by GC- or LC-MS