Learn More
Apexbio Technology LLC Guanabenz Acetate 23256-50-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000089741
Encompass_Preferred
Guanabenz Acetate (CAS 23256-50-0) is a small molecule that acts as a selective agonist of the 2-adrenergic receptor subtypes It exhibits notable affinity for the 2a-adrenergic receptor (pEC50 8 25) with moderate activity at the 2b-adrenergic receptor (pEC50 7 01) and lower potency at the 2c-adrenergic receptor (pEC50 5) Through activation of these receptors Guanabenz Acetate modulates adrenergic neurotransmission making it a valuable tool for investigating the physiological and pharmacological functions of 2-adrenergic signaling in biomedical research
Catalog No. 50-000-89741 Supplier Apexbio Technology LLC Supplier No. B13355.1
Guanabenz Acetate (CAS 23256-50-0) is a small molecule that acts as a selective agonist of the 2-adrenergic receptor subtypes It exhibits notable affinity for the 2a-adrenergic receptor (pEC50 8 25) with moderate activity at the 2b-adrenergic receptor (pEC50 7 01) and lower potency at the 2c-adrenergic receptor (pEC50 5) Through activation of these receptors Guanabenz Acetate modulates adrenergic neurotransmission making it a valuable tool for investigating the physiological and pharmacological functions of 2-adrenergic signaling in biomedical research