Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guanabenz acetate, Tocris Bioscience™

Catalog No. 0885100
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
08-851-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 08-851-00 Supplier R&D Systems Supplier No. 0885/100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

α2 agonist. Also I2 selective ligand

α2-adrenergic agonist and IGRS (imidazoline I2 binding site) selective ligand.

Chemical Identifiers

CAS 23256-50-0
Molecular Formula C10H12Cl2N4O2
Molecular Weight (g/mol) 291.132
InChI Key MCSPBPXATWBACD-UHFFFAOYSA-N
Synonym guanabenz wy-8678 acetate, 2,6-dichlorobenzylidene-amino guanidine, acetic acid; 2-2,6-dichlorophenyl methylideneamino guanidine, n-2,6-dichlorophenyl methylidene amino guanidine; acetic acid
PubChem CID 6852405
IUPAC Name acetic acid;2-[(2,6-dichlorophenyl)methylideneamino]guanidine
SMILES CC(=O)O.C1=CC(=C(C(=C1)Cl)C=NN=C(N)N)Cl

Specifications

Quantity 100 mg
Formula Weight 291.14
Percent Purity >98%
Chemical Name or Material Guanabenz acetate
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.